Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cis-3-hexenyl hexanoate | 31501-11-8 | MFCD00036552 | 98.0% | 198.30 | C12H22O2 | 500 MG
SDP

Catalog No. 5000303165
Encompass_Preferred

Cis-3-hexenyl hexanoate is a fatty acid ester (n-caproic acid cis-3-hexen-1-yl ester) used as a reference standard and in flavor and fragrance research. It is supplied with analytical documentation and storage guidance, and is available in multiple pack sizes and solution formats.

  • High purity (98.0%).
  • Molecular formula C12H22O2 and molecular weight 198.30.
  • Supplied with COA, SDS, HNMR, and MS documentation.
  • Available as solids and as 10 mM solutions in DMSO across multiple pack sizes.
  • Storage: store at 4°C protected from light; in solvent store at -20°C short term or -80°C for long term.

Catalog No. 50-003-03165 Supplier Medchemexpress LLC Supplier No. HYW014933500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cis-3-hexenyl hexanoate is a fatty acid ester (n-caproic acid cis-3-hexen-1-yl ester) used as a reference standard and in flavor and fragrance research. It is supplied with analytical documentation and storage guidance, and is available in multiple pack sizes and solution formats.

  • High purity (98.0%).
  • Molecular formula C12H22O2 and molecular weight 198.30.
  • Supplied with COA, SDS, HNMR, and MS documentation.
  • Available as solids and as 10 mM solutions in DMSO across multiple pack sizes.
  • Storage: store at 4°C protected from light; in solvent store at -20°C short term or -80°C for long term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.