Learn More
Medchemexpress LLC Cis-3-hexenyl hexanoate | 31501-11-8 | MFCD00036552 | 98.0% | 198.30 g/mol | C12H22O2 | 1 ML
SDP
Catalog No. 5000303349
Encompass_Preferred
cis-3-Hexenyl hexanoate is the cis-3-hexen-1-yl ester of hexanoic (caproic) acid, a volatile flavor and fragrance compound commonly used as a chemical standard and research reagent. It is provided as a solution formulated for laboratory use and intended for olfactory studies, flavor chemistry, and analytical testing.
  • used as a flavor and fragrance reference compound.
  • supplied as a 10 mM solution in DMSO for ready-to-use applications.
  • high purity (98.0%) suitable for analytical and research use.
  • molecular formula C12H22O2; molecular weight 198.30 g/mol.
  • commonly applied in GC/MS analysis and olfactory research workflows.
Catalog No. 50-003-03349 Supplier Medchemexpress LLC Supplier No. HYW01493310MM/1ML
cis-3-Hexenyl hexanoate is the cis-3-hexen-1-yl ester of hexanoic (caproic) acid, a volatile flavor and fragrance compound commonly used as a chemical standard and research reagent. It is provided as a solution formulated for laboratory use and intended for olfactory studies, flavor chemistry, and analytical testing.
  • used as a flavor and fragrance reference compound.
  • supplied as a 10 mM solution in DMSO for ready-to-use applications.
  • high purity (98.0%) suitable for analytical and research use.
  • molecular formula C12H22O2; molecular weight 198.30 g/mol.
  • commonly applied in GC/MS analysis and olfactory research workflows.