Learn More
Medchemexpress LLC Cis-3-hexenyl hexanoate | 31501-11-8 | MFCD00036552 | 98.0% | 198.30 g/mol | C12H22O2 | 100 MG
SDP
Catalog No. 5000303146
Encompass_Preferred
Cis-3-hexenyl hexanoate is a volatile, fruity ester (n-caproic acid cis-3-hexen-1-yl ester) used as a research reagent and as a flavor and fragrance building block. It has been reported as a natural constituent of Feijoa (Acca sellowiana) and is supplied in solution and bulk formats for laboratory applications. Intended for research use only.
  • High purity (98.0%) appropriate for analytical and preparative work.
  • Fruity, green aroma useful as a flavor and fragrance building block.
  • Available in solution and multiple pack sizes for flexible use.
  • Stable when stored under recommended conditions; solvent storage guidance provided.
  • Molecular formula C12H22O2; molecular weight 198.30 g/mol.
Catalog No. 50-003-03146 Supplier Medchemexpress LLC Supplier No. HYW014933100MG
Cis-3-hexenyl hexanoate is a volatile, fruity ester (n-caproic acid cis-3-hexen-1-yl ester) used as a research reagent and as a flavor and fragrance building block. It has been reported as a natural constituent of Feijoa (Acca sellowiana) and is supplied in solution and bulk formats for laboratory applications. Intended for research use only.
  • High purity (98.0%) appropriate for analytical and preparative work.
  • Fruity, green aroma useful as a flavor and fragrance building block.
  • Available in solution and multiple pack sizes for flexible use.
  • Stable when stored under recommended conditions; solvent storage guidance provided.
  • Molecular formula C12H22O2; molecular weight 198.30 g/mol.