Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-3-Hexenyl Pyruvate 95.0+%, TCI America™
SDP

Catalog No. P089425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P089425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P089425ML Supplier TCI America Supplier No. P089425ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 68133-76-6
Molecular Formula C9H14O3
Molecular Weight (g/mol) 170.21
MDL Number MFCD00036527
InChI Key LKNXTZXOBHAYSR-PLNGDYQASA-N
Synonym Pyruvic Acid cis-3-Hexenyl Ester
PubChem CID 5363291
IUPAC Name (3Z)-hex-3-en-1-yl 2-oxopropanoate
SMILES CC\C=C/CCOC(=O)C(C)=O

Specifications

Color Yellow
Boiling Point 98°C
Quantity 25 mL
Formula Weight 170.21
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material cis-3-Hexenyl Pyruvate
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.