Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-4-Amino-N-Boc-L-proline methyl ester hydrochloride, 97%

Catalog No. p-7035029
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52731MD 250 mg
AAH5273103 1 g
AAH5273106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH52731MD Supplier Thermo Scientific Chemicals Supplier No. H52731MD
Only null left
Add to Cart

CAS: 171110-72-8 | C11H21ClN2O4 | 280.75 g/mol

cis-4-Amino-N-Boc-L-proline methyl ester hydrochloride is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-4-Amino-N-Boc-L-proline methyl ester hydrochloride is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 171110-72-8
Molecular Formula C11H21ClN2O4
Molecular Weight (g/mol) 280.75
MDL Number MFCD03787927
InChI Key KFYCQKLSGMAVQH-UHFFFAOYNA-N
Synonym methyl 2s,4s-1-boc-4-aminopyrrolidine-2-carboxylate hydrochloride, 2s,4s-1-tert-butyl 2-methyl 4-aminopyrrolidine-1,2-dicarboxylate hydrochloride, n-boc-cis-4-amino-l-proline methyl ester hydrochloride, 2s,4s-1-boc-4-amino pyrrolidine-2-carboxylic acid methylester-hcl, 1-tert-butyl 2-methyl 2s,4s-4-aminopyrrolidine-1,2-dicarboxylate hydrochloride, n-boc-cis-4-amino-l-proline methyl ester hydrochloride salt, 1-tert-butyl 2-methyl cis-4-aminopyrrolidine-1,2-dicarboxylate hydrochloride, 2s,4s-1-boc-4-amino pyrrolidine-2-carboxylic acid methylester hydrochloride, 4-aminopyrrolidine-1,2-dicarboxylatehydrochloride, s,s-1-boc-4-aminoproline methyl ester hydrochloride
PubChem CID 17998957
SMILES Cl.COC(=O)C1CC(N)CN1C(=O)OC(C)(C)C

Specifications

Melting Point ∽185°C (decomposition)
Quantity 250 mg
Optical Rotation −40° (c=2 in Methanol)
IUPAC Name 1-tert-butyl 2-methyl 4-aminopyrrolidine-1,2-dicarboxylate hydrochloride
Formula Weight 280.75
Percent Purity 97%
Chemical Name or Material cis-4-Amino-N-Boc-L-proline methyl ester hydrochloride
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.