Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-4-(Boc-amino)cyclohexanecarboxaldehyde, 97%

Catalog No. AAH62871MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH62871MD 250 mg
AAH6287103 1 g
AAH6287106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH62871MD Supplier Thermo Scientific Chemicals Supplier No. H62871MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 181308-56-5 | C12H21NO3 | 227.304 g/mol

cis-4-(Boc-amino)cyclohexanecarboxaldehyde is used as a intermediate for chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-4-(Boc-amino)cyclohexanecarboxaldehyde is used as a intermediate for chemical research.

Notes
Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 181308-56-5
Molecular Formula C12H21NO3
Molecular Weight (g/mol) 227.304
MDL Number MFCD03844599
InChI Key GPDBIGSFXXKWQR-UHFFFAOYSA-N
Synonym tert-butyl trans-4-formylcyclohexylcarbamate, tert-butyl cis-4-formylcyclohexylcarbamate, trans-boc-4-aminocyclohexanecarbaldehyde, tert-butyl n-4-formylcyclohexyl carbamate, trans-4-formyl-cyclohexyl-carbamic acid tert-butyl ester, tert-butyl trans-4-formylcyclohexyl carbamate, tert-butyl 4-formylcyclohexyl carbamate, trans-tert-butyl 4-formylcyclohexyl carbamate, tert-butyl 1r,4r-4-formylcyclohexylcarbamate, cis-4-boc-amino cyclohexanecarboxaldehyde
PubChem CID 11020615
IUPAC Name tert-butyl N-(4-formylcyclohexyl)carbamate
SMILES CC(C)(C)OC(=O)NC1CCC(CC1)C=O

Specifications

Quantity 250 mg
Formula Weight 227.3
Percent Purity 97%
Chemical Name or Material cis-4-(Boc-amino)cyclohexanecarboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.