Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-4-Cyanocyclohexylamine hydrochloride, 97%, Thermo Scientific™

Catalog No. AAH6229203
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6229203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6229203 Supplier Thermo Scientific Chemicals Supplier No. H6229203

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
cis-4-Cyanocyclohexylamine hydrochloride is used in chemical research.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1387445-51-3
Molecular Formula C7H13ClN2
Molecular Weight (g/mol) 160.645
MDL Number MFCD22378474
InChI Key BTLVSLNZSMIPCW-UHFFFAOYSA-N
Synonym trans-4-aminocyclohexanecarbonitrile hydrochloride, cis-4-aminocyclohexanecarbonitrile hydrochloride, 4-amino-cyclohexanecarbonitrile hydrochloride, 4-aminocyclohexanecarbonitrile hydrochloride, 4-aminocyclohexane-1-carbonitrile hydrochloride, c7h12n2.hcl, 1s,4s-4-aminocyclohexanecarbonitrile hydrochloride, 1r,4r-4-aminocyclohexane-1-carbonitrile hydrochloride, 1s,4s-4-aminocyclohexane-1-carbonitrile hydrochloride, cyclohexanecarbonitrile, 4-amino-, monohydrochloride, trans
PubChem CID 53407876
IUPAC Name 4-aminocyclohexane-1-carbonitrile;hydrochloride
SMILES C1CC(CCC1C#N)N.Cl

Specifications

Quantity 1 g
UN Number UN3439
Formula Weight 160.64
Percent Purity 97%
Chemical Name or Material cis-4-Cyanocyclohexylamine hydrochloride
Hazard Category Warning
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.