Learn More
Medchemexpress LLC cis-4-Decen-1-ol 25g
SDP
Catalog No. 5000206395
Encompass_Preferred
Cis-4-Decen-1-ol is an organic compound belonging to the group of alcohols It has a floral scent and is commonly found in a variety of foods including fruits and vegetables Cis-4-Decen-1-ol has several applications in the flavor and fragrance industry especially as a fragrance agent in products such as perfumes colognes and air fresheners In addition it has potential utility in inhibiting inflammation-related diseases and cancer
Catalog No. 50-002-06395 Supplier Medchemexpress LLC Supplier No. 25GHYW127518
Cis-4-Decen-1-ol is an organic compound belonging to the group of alcohols It has a floral scent and is commonly found in a variety of foods including fruits and vegetables Cis-4-Decen-1-ol has several applications in the flavor and fragrance industry especially as a fragrance agent in products such as perfumes colognes and air fresheners In addition it has potential utility in inhibiting inflammation-related diseases and cancer