Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC cis-4-hydroxy-L-proline hydrochloride | 441067-49-8 | MFCD09953187 | 97.0% | 167.59 g/mol | C5H10ClNO3 | 5 G
SDP

Catalog No. 5000203068
Encompass_Preferred

cis-4-hydroxy-L-proline hydrochloride is a proline analog used in research as a collagen production inhibitor and as a building block in linker and medicinal chemistry. It is supplied as a white to light-yellow solid with reported purity and defined storage conditions.

  • White to light-yellow solid.
  • Used as a collagen production inhibitor and building block in medicinal chemistry.
  • Molecular weight 167.59 g/mol.
  • Chemical formula C5H10ClNO3.
  • Purity 97.0% (NMR).
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Available in small-scale laboratory quantities (for example, 5 g).

Catalog No. 50-002-03068 Supplier Medchemexpress LLC Supplier No. HYW0192135G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

cis-4-hydroxy-L-proline hydrochloride is a proline analog used in research as a collagen production inhibitor and as a building block in linker and medicinal chemistry. It is supplied as a white to light-yellow solid with reported purity and defined storage conditions.

  • White to light-yellow solid.
  • Used as a collagen production inhibitor and building block in medicinal chemistry.
  • Molecular weight 167.59 g/mol.
  • Chemical formula C5H10ClNO3.
  • Purity 97.0% (NMR).
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Available in small-scale laboratory quantities (for example, 5 g).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.