Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-4-(Hydroxymethyl)cyclohexanecarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. H12425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12425G 5 g
H124225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H12425G Supplier TCI America Supplier No. H12425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 73094-35-6
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.197
InChI Key VQMIUUBKKPIDBN-UHFFFAOYSA-N
PubChem CID 202819
IUPAC Name 4-(hydroxymethyl)cyclohexane-1-carboxylic acid
SMILES C1CC(CCC1CO)C(=O)O

Specifications

Melting Point 120°C
Color White-Yellow
Quantity 5 g
Formula Weight 158.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material cis-4-(Hydroxymethyl)cyclohexanecarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.