Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-4-Methylcyclohexylamine 98.0+%, TCI America™
SDP

Catalog No. M2949200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2949200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M2949200MG Supplier TCI America Supplier No. M2949200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2523-56-0
Molecular Formula C7H15N
Molecular Weight (g/mol) 113.204
MDL Number MFCD13563117
InChI Key KSMVBYPXNKCPAJ-UHFFFAOYSA-N
PubChem CID 80604
IUPAC Name 4-methylcyclohexan-1-amine
SMILES CC1CCC(CC1)N

Specifications

Color Yellow
Boiling Point 154°C
Quantity 200 mg
UN Number 2734
Formula Weight 113.20
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material cis-4-Methylcyclohexylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.