Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-5,8,11,14,17-Eicosapentaenoic acid, ≥98.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012361 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2361 500 mg
11-101-2360 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-2361 Supplier MilliporeSigma Supelco Supplier No. 44864500MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 10417-94-4
Density 0.943 g/mL (at 25°C)
Flash Point 93°C
Physical Form Neat
Linear Formula CH3(CH2 CH=CH)5(CH2)3 CO2 H
MDL Number MFCD00065716
Melting Point -54°C to -53°C
Synonym EPA; Timnodonic acid
Type Analytical Standard
Quantity 500 mg
Formula Weight 302.45
Percent Purity ≥98.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.