Learn More
Medchemexpress LLC cis-5-Dodecenoic acid | 2430-94-6 | 1 ML
SDP
Catalog No. 5000419216
Encompass_Preferred
cis-5-Dodecenoic acid is an inhibitor of COX-I and COX-II, exhibiting anti-inflammatory activity. It functions by reducing prostaglandin synthesis through the inhibition of COX enzyme activity and participates in the fatty acid -β oxidative metabolic pathway. The metabolic rate of cis-5-Dodecenoic acid is notably lower than that of saturated fatty acids.
  • Can be used in research of anti-inflammation, fatty acid metabolism mechanisms, and related physiological and pathological processes.
  • Purity is 99.76%.
  • Exhibits inhibitory activities on COX-I and COX-II (35% and 32% respectively at 100 μg/mL).
  • Involved in metabolism by conversion to cis-5-dodecenoyl-CoA.
  • Dehydrogenation rate of cis-5-dodecenoyl-CoA, after β-oxidation, is significantly lower (10-44%) compared to saturated acyl-CoA.
Catalog No. 50-004-19216 Supplier Medchemexpress LLC Supplier No. HYN846910MM/1ML
Add to Cart
Add to Cart
cis-5-Dodecenoic acid is an inhibitor of COX-I and COX-II, exhibiting anti-inflammatory activity. It functions by reducing prostaglandin synthesis through the inhibition of COX enzyme activity and participates in the fatty acid -β oxidative metabolic pathway. The metabolic rate of cis-5-Dodecenoic acid is notably lower than that of saturated fatty acids.
  • Can be used in research of anti-inflammation, fatty acid metabolism mechanisms, and related physiological and pathological processes.
  • Purity is 99.76%.
  • Exhibits inhibitory activities on COX-I and COX-II (35% and 32% respectively at 100 μg/mL).
  • Involved in metabolism by conversion to cis-5-dodecenoyl-CoA.
  • Dehydrogenation rate of cis-5-dodecenoyl-CoA, after β-oxidation, is significantly lower (10-44%) compared to saturated acyl-CoA.