Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC cis-5-Dodecenoic acid | 2430-94-6 | 25 MG
SDP

Catalog No. 5000421474
Encompass_Preferred

cis-5-Dodecenoic acid is an inhibitor of COX-I and COX-II with anti-inflammatory activity. It reduces prostaglandin synthesis by inhibiting COX enzyme activity and is involved in the fatty acid -β oxidative metabolic pathway. Its metabolic rate is significantly lower than that of saturated fatty acids, making it suitable for research into anti-inflammation, fatty acid metabolism mechanisms, and related physiological and pathological processes.

  • Inhibitor of COX-I and COX-II
  • Reduces prostaglandin synthesis
  • Involved in the fatty acid -β oxidative metabolic pathway
  • Metabolic rate is significantly lower than that of saturated fatty acids
  • Purity: 99.76%
  • Appearance: Liquid, colorless to light yellow
  • Possesses anti-inflammatory activity
  • Can be used in research for anti-inflammation, fatty acid metabolism mechanisms, and related physiological and pathological processes

Catalog No. 50-004-21474 Supplier Medchemexpress LLC Supplier No. 25MGHYN8469
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

cis-5-Dodecenoic acid is an inhibitor of COX-I and COX-II with anti-inflammatory activity. It reduces prostaglandin synthesis by inhibiting COX enzyme activity and is involved in the fatty acid -β oxidative metabolic pathway. Its metabolic rate is significantly lower than that of saturated fatty acids, making it suitable for research into anti-inflammation, fatty acid metabolism mechanisms, and related physiological and pathological processes.

  • Inhibitor of COX-I and COX-II
  • Reduces prostaglandin synthesis
  • Involved in the fatty acid -β oxidative metabolic pathway
  • Metabolic rate is significantly lower than that of saturated fatty acids
  • Purity: 99.76%
  • Appearance: Liquid, colorless to light yellow
  • Possesses anti-inflammatory activity
  • Can be used in research for anti-inflammation, fatty acid metabolism mechanisms, and related physiological and pathological processes

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.