Alfa Aesar™ cis-5-Norbornene-endo-2,3-dicarboxylic acid, 98%

Manufacturer: Alfa Aesar™ H5340314

Quantity 25g

 View more versions of this product

Catalog No. AAH5340314

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3853-88-1
    Molecular Formula C9H10O4
    Formula Weight 182.18
    MDL Number MFCD00003735
    Synonym cis-endo-Bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic acid
    Chemical Name or Material cis-5-Norbornene-endo-2,3-dicarboxylic acid

    Specifications

    Formula Weight 182.18
    MDL Number MFCD00003735
    Quantity 25g
    Molecular Formula C9H10O4
    CAS 3853-88-1
    Synonym cis-endo-Bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic acid
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Melting Point 175°C
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material cis-5-Norbornene-endo-2,3-dicarboxylic acid
    Assay 98%

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit