Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-5-Octen-1-ol 90.0+%, TCI America™
SDP

Catalog No. O025010ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O025010ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O025010ML Supplier TCI America Supplier No. O025010ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 64275-73-6
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.215
MDL Number MFCD00015569
InChI Key VDHRTASWKDTLER-ONEGZZNKSA-N
Synonym oct-5-en-1-ol, e-oct-5-en-1-ol, 5-octen-1-ol, 5e-oct-5-en-1-ol
PubChem CID 6008958
IUPAC Name (E)-oct-5-en-1-ol
SMILES CCC=CCCCCO

Specifications

Color Yellow
Boiling Point 97°C
Quantity 10 mL
Formula Weight 128.22
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material cis-5-Octen-1-ol
EINECSNumber (2)-2393
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.