Learn More
Cayman Chemical cis-6-Hexadecenoic Acid, Purity: ≥99%, CAS: 17004-51-2, 10 mg
SDP
Catalog No. 502893185
Encompass_Preferred
A monounsaturated fatty acid; one of the primary fatty acids in human skin; levels are increased in isolated sebum from the face and back of patients with acne; levels are decreased in the non-lesional skin and isolated sebum of atopic dermatitis patients; active against S. aureus in vitro at 5 µg/ml at pH 5.5
Catalog No. 50-289-3185 Supplier Cayman Chemical Supplier No. 900184510 MG
A monounsaturated fatty acid; one of the primary fatty acids in human skin; levels are increased in isolated sebum from the face and back of patients with acne; levels are decreased in the non-lesional skin and isolated sebum of atopic dermatitis patients; active against S. aureus in vitro at 5 µg/ml at pH 5.5