Learn More
Apexbio Technology LLC cis-6-Octadecenoic acid 593-39-5 25mg
SDP
Catalog No. 5000095956
Encompass_Preferred
cis-6-Octadecenoic acid (CAS 593-39-5) is a monounsaturated fatty acid comprising 18 carbon atoms with a cis double bond at the 6 position It is present in certain plant seed oils and some animal fats though less commonly than other octadecenoic acid isomers Research has explored its influence on lipid metabolism membrane fluidity and cellular signaling pathways reflecting the broader roles of unsaturated fatty acids in physiological and pathological processes cis-6-Octadecenoic acid serves as a valuable tool for investigating mechanisms related to fatty acid function lipid homeostasis and membrane dynamics in biomedical studies
Catalog No. 50-000-95956 Supplier Apexbio Technology LLC Supplier No. A996225
cis-6-Octadecenoic acid (CAS 593-39-5) is a monounsaturated fatty acid comprising 18 carbon atoms with a cis double bond at the 6 position It is present in certain plant seed oils and some animal fats though less commonly than other octadecenoic acid isomers Research has explored its influence on lipid metabolism membrane fluidity and cellular signaling pathways reflecting the broader roles of unsaturated fatty acids in physiological and pathological processes cis-6-Octadecenoic acid serves as a valuable tool for investigating mechanisms related to fatty acid function lipid homeostasis and membrane dynamics in biomedical studies