Learn More
Cayman Chemical cis-7-Hexadecenoic Acid, 2416-19-5, 5 mg
SDP
Supplier:  Cayman Chemical 100072905
Encompass_Preferred
Molecular Formula C16H30O2, Purity ≥98%, Formula Weight 254.4, Synonyms: 7Z-hexadecenoic acid; Mono-unsaturated fatty acids are components of the cellular membranes of autotrophic bacteria. The specific composition and abundance of membrane fatty acids can be used to identify specific genera of bacterial populations in natural environments (e.g., mining lakes, etc.).{13903} cis-7-Hexadecenoic acid has been isolated from autotrophic bacterial cultures associated with the accumulation of sulfate in biofilters. This indicates they originate from a sulfide-oxidizing autotrophic organism. To date, however, this fatty acid has only been detected in strains of the genus Nitrospira, which are nitrite oxidizing autotrophic bacteria.
Catalog No. NC1530658