Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-8,11,14-Eicosatrienoic acid, ≥98.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018589 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8589 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8589 Supplier MilliporeSigma Supelco Supplier No. 6915450MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 1783-84-2
Flash Point 62°C - closed cup
Physical Form Viscous Liquid
Linear Formula CH3(CH2)3(CH2 CH=CH)3(CH2)6 CO2 H
MDL Number MFCD00065721
Synonym Homo-gamma-linolenic acid
Type Analytical Standard
Quantity 50 mg
Formula Weight 306.48
Percent Purity ≥98.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.