Learn More
Cayman Chemical cis-9,10-Epoxyoctadecanoic Acid, Purity: ≥95%, CAS: 24560-98-3, 10 mg
SDP
Catalog No. 502911416
Encompass_Preferred
An epoxy fatty acid; has been found in the fungus P. carinii isolated from the lungs of rats in a model of P. carinii-induced pneumonia; plasma levels are decreased in patients with impaired liver function
Catalog No. 50-291-1416 Supplier Cayman Chemical Supplier No. 3644210 MG
An epoxy fatty acid; has been found in the fungus P. carinii isolated from the lungs of rats in a model of P. carinii-induced pneumonia; plasma levels are decreased in patients with impaired liver function