Learn More
Medchemexpress LLC HY-129554 5mg Medchemexpress, cis-​9,​10-​Epoxystearic acid CAS:24560-98-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1295545MG
Encompass_Preferred
Medchemexpress, HY-129554 5mg cis-​9,​10-​Epoxystearic acid CAS:24560-98-3 cis-9,10-Epoxystearic acid (cis-9,10-Epoxyoctadecanoic acid) is an endogenous constituent in human blood and urine. cis-9,10-Epoxystearic acid (cis-9,10-Epoxyoctadecanoic acid) can be produced from oleic acid by enzymic and non-enzymic epoxidation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9811