Learn More
Abcam cis-ACBD (1-Aminocyclobutane-1,3-dicarbo x ylic acid), L-glutamate uptake inhibitor, 50MG
Catalog No. 502704944
Encompass_Preferred
MW 159.14 Da. Selective, competitive L-glutamate uptake inhibitor (IC₅₀ = 30 μM). Increases basal and potassium evoked D-aspartate release.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4944 Supplier Abcam Supplier No. AB14612850MG
MW 159.14 Da. Selective, competitive L-glutamate uptake inhibitor (IC₅₀ = 30 μM). Increases basal and potassium evoked D-aspartate release.
The product is subject to the following: Abcam Restricted Use Statement