Learn More
Medchemexpress LLC Cis-aconitic acid | 585-84-2 | MFCD00063184 | 99.3% | 174.11 g/mol | C6H6O6 | 5 MG
SDP
Catalog No. 5000301048
Encompass_Preferred
(Z)-Aconitic acid is a tricarboxylic organic acid used as a research reagent in biochemical and pharmacological studies. It has reported activity as a glutamate decarboxylase inhibitor, reduces IκB-α phosphorylation, and has been used in models of antigen-induced arthritis and monosodium urate-induced gout. Available in solid form and as DMSO solutions for in vitro and in vivo applications.
  • High purity suitable for research applications.
  • Available in small solid quantities and DMSO solutions.
  • Used in biochemical assays and pharmacological studies.
  • Reported to inhibit glutamate decarboxylase activity.
  • Documented activity in inflammatory disease models.
Catalog No. 50-003-01048 Supplier Medchemexpress LLC Supplier No. HYW016814R5MG
(Z)-Aconitic acid is a tricarboxylic organic acid used as a research reagent in biochemical and pharmacological studies. It has reported activity as a glutamate decarboxylase inhibitor, reduces IκB-α phosphorylation, and has been used in models of antigen-induced arthritis and monosodium urate-induced gout. Available in solid form and as DMSO solutions for in vitro and in vivo applications.
  • High purity suitable for research applications.
  • Available in small solid quantities and DMSO solutions.
  • Used in biochemical assays and pharmacological studies.
  • Reported to inhibit glutamate decarboxylase activity.
  • Documented activity in inflammatory disease models.