Learn More
Selleck Chemical LLC cis-Bac430 E4958-100MG
SDP
Catalog No. 5000041768
Encompass_Preferred
Cis-Bac430 plays a role in mediating the movement of anti-tumor CD8 cells from the periphery to the tumor site thus enhancing the anti-tumor effect of an immune checkpoint inhibitor
Catalog No. 50-000-41768 Supplier Selleck Chemical LLC Supplier No. E4958100MG
Cis-Bac430 plays a role in mediating the movement of anti-tumor CD8 cells from the periphery to the tumor site thus enhancing the anti-tumor effect of an immune checkpoint inhibitor