Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cis,Cis-5,8-Tetradecanedienoyl-L-carnitine Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111022439 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-2439 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-2439 Supplier MilliporeSigma Supelco Supplier No. 1918810MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 1469901-01-6
Flash Point Not applicable
Molecular Formula C21H37NO4
Linear Formula C21H37NO4
Quantity 10 mg
UN Number NONH for all modes of transport
Synonym (2 R)-3-Carboxy-N,N,N-trimethyl-2-[[(5 Z,8 Z)-1-oxo-5,8-tetradecadien-1-yl]oxy]-1-propanaminium inner salt
Optical Rotation [α]/D -18°C ± 2°C = 0.1 in. methanol
Formula Weight 367.52
Percent Purity ≥95% (LC/MS)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.