Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis,cis-Muconic acid, 98%

Catalog No. AC297760100
Change view
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC297760100 10 g Glass bottle
AC297760025 2.5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC297760100 Supplier Thermo Scientific Chemicals Supplier No. 297760100
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1119-72-8 | C6H6O4 | 142.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1119-72-8
Molecular Formula C6H6O4
Molecular Weight (g/mol) 142.11
MDL Number MFCD00085266
InChI Key TXXHDPDFNKHHGW-ZPUQHVIOSA-N
Synonym cis,cis-muconic acid, 2z,4z-hexa-2,4-dienedioic acid, cis,cis-muconate, unii-2g78tuq51g, cis,cis-2,4-hexadienedioic acid, 2,4-hexadienedioic acid, z,z, 2z,4z-hexadienedioic acid, cis,cis-hexadienedioate, z,z-2,4-hexadienedioic acid, 2,4-hexadienedioic acid
PubChem CID 5280518
ChEBI CHEBI:16508
SMILES OC(=O)\C=C\C=C\C(O)=O

Specifications

Melting Point 194°C to 195°C
Color Beige-Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula HO2CCH=CHCH=CHCO2H
Quantity 10 g
Merck Index 15, 6385
Solubility Information Solubility in water: freely soluble in boiling water. Other solubilities: sparingly soluble in diethyl ether
IUPAC Name (2Z,4Z)-hexa-2,4-dienedioic acid
Formula Weight 142.11
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material cis, cis-Muconic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.