Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-Decahydronaphthalene 98.0+%, TCI America™
SDP

Catalog No. D0009500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0009500ML 500 mL
D000925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0009500ML Supplier TCI America Supplier No. D0009500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 493-01-6
Molecular Formula C10H18
Molecular Weight (g/mol) 138.25
MDL Number MFCD00004130,MFCD00064189,MFCD00064191
InChI Key NNBZCPXTIHJBJL-UHFFFAOYSA-N
Synonym decalin, decahydronaphthalene, cis-decahydronaphthalene, trans-decahydronaphthalene, cis-decalin, dekalin, naphthalene, decahydro, perhydronaphthalene, trans-decalin, naphthalane
PubChem CID 7044
ChEBI CHEBI:38853
IUPAC Name decahydronaphthalene
SMILES C1CCC2CCCCC2C1

Specifications

Color Colorless
Boiling Point 193°C
Quantity 500 mL
UN Number 1147
Formula Weight 138.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material cis-Decahydronaphthalene
EINECSNumber (4)-0575
RTECSNumber QJ3150000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.