Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cis-Dichlorobis(Diethyl Sulfide)Platinum(II)

Catalog No. 4328491G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-284-91G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-284-91G Supplier Sigma Aldrich Supplier No. 4328491G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Cis-Dichlorobis(Diethyl Sulfide)Platinum(II)

Specifications

CAS 56210-72-1
Density 0.987 g/mL at 25°C
Boiling Point 86°C (0.05 mmHg, lit.)
Molecular Formula C16H19N
Linear Formula [C6H5CH(CH3)]2NH
Quantity 1 g
MDL Number MFCD00243087
Synonym cis-Dichlorobis(1,1'-thiobisethane)platinum; cis-Bis(diethyl sulfide)platinum(II) chloride
Molecular Weight (g/mol) 225.33 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.