Learn More
Strem, An Ascensus Company CAS# 15692-07-6. 500mg. cis-Dichlorobis(triethylphosphine)platinum(II), 99%. MFCD00058821
SDP
Supplier:  Strem, An Ascensus Company 937820500MG
Encompass
CAS# 15692-07-6. 500mg. cis-Dichlorobis(triethylphosphine)platinum(II), 99%. MFCD00058821. Molecular Weight: 502.32. Molecular Formula: PtCl2[(C2H5)3P]2. Color/form: white to off-white pwdr. Strem# 93-7820. http://www.strem.com/catalog/v/93-7820/
Catalog No. 50-901-16370