Learn More
Strem, An Ascensus Company CAS# 15663-27-1. 5g. cis-Dichlorodiammine platinum(II), 99% CISPLATIN. MFCD00011623
SDP
Catalog No. 5090113219
Encompass
CAS# 15663-27-1. 5g. cis-Dichlorodiammine platinum(II), 99% CISPLATIN. MFCD00011623. Molecular Weight: 300.06. Molecular Formula: cis-Pt(NH3)2Cl2. Color/form: yellow to orange xtl. Strem# 78-0450. http://www.strem.com/catalog/v/78-0450/
Catalog No. 50-901-13219 Supplier Strem, An Ascensus Company Supplier No. 7804505G
CAS# 15663-27-1. 5g. cis-Dichlorodiammine platinum(II), 99% CISPLATIN. MFCD00011623. Molecular Weight: 300.06. Molecular Formula: cis-Pt(NH3)2Cl2. Color/form: yellow to orange xtl. Strem# 78-0450. http://www.strem.com/catalog/v/78-0450/