Learn More
Cayman Chemical cis-Entacapone, Purity: ≥98%, CAS: 145195-63-7, 5 mg
SDP
Catalog No. 502938225
Encompass_Preferred
A metabolite of entacapone; a potential impurity found in commercial preparations of entacapone; a degradant of entacapone formed by UV light exposure
Catalog No. 50-293-8225 Supplier Cayman Chemical Supplier No. 280465 MG
A metabolite of entacapone; a potential impurity found in commercial preparations of entacapone; a degradant of entacapone formed by UV light exposure