Learn More
Cayman Chemical -cIs-EpoxIconazole-d4 1mg
SDP
Catalog No. 502932666
Encompass_Preferred
An internal standard for the quantification of cis-epoxiconazole by GC- or LC-MS
Catalog No. 50-293-2666 Supplier Cayman Chemical Supplier No. 267711 MG
Add to Cart
Add to Cart
An internal standard for the quantification of cis-epoxiconazole by GC- or LC-MS