Learn More
Cayman Chemical -cIs-EpoxIconazole-d4 500ug
SDP
Catalog No. 502935208
Encompass_Preferred
An internal standard for the quantification of cis-epoxiconazole by GC- or LC-MS
Catalog No. 50-293-5208 Supplier Cayman Chemical Supplier No. 26771500 UG
An internal standard for the quantification of cis-epoxiconazole by GC- or LC-MS