Learn More
Medchemexpress LLC Cis-epoxysuccinic acid | 16533-72-5 | MFCD00191645 | >97.0% | 132.07 | C4H4O5 | 100MG
SDP
Catalog No. 5000238025
Encompass_Preferred
cis-Epoxysuccinic acid is a research-grade small molecule that acts as a succinate receptor (SUCNR1/GPR91) agonist. It is supplied as a white to off-white solid with molecular formula C4H4O5 and molecular weight 132.07, typically offered at >97.0% purity for laboratory use. Reported activity includes inhibition of cAMP with an EC50 of 2.7 μM. Store protected from light at 4°C; in solvent keep at -80°C for long-term storage.
Catalog No. 50-002-38025 Supplier Medchemexpress LLC Supplier No. HY125791100MG
cis-Epoxysuccinic acid is a research-grade small molecule that acts as a succinate receptor (SUCNR1/GPR91) agonist. It is supplied as a white to off-white solid with molecular formula C4H4O5 and molecular weight 132.07, typically offered at >97.0% purity for laboratory use. Reported activity includes inhibition of cAMP with an EC50 of 2.7 μM. Store protected from light at 4°C; in solvent keep at -80°C for long-term storage.