Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cis-epoxysuccinic acid | 16533-72-5 | MFCD00191645 | 98.0% | 132.07 g/mol | C4H4O5 | 10MM 1ML
SDP

Catalog No. 5000236405
Encompass_Preferred

cis-Epoxysuccinic acid is a small-molecule agonist of the succinate receptor (SUCNR1) used in receptor pharmacology, cell signaling research, and biochemical assays. It is supplied as a white to off-white solid and as a solution (commonly 10 mM in DMSO, 1 mL); handle according to the safety data sheet and storage recommendations.

  • Purity 98.0%.
  • Molecular weight 132.07 g/mol.
  • Chemical formula C4H4O5.
  • Available as 10 mM solution in DMSO (1 mL) and solid sizes 100 mg-5 g.
  • Appearance white to off-white solid.
  • Intended for SUCNR1 pharmacology and biochemical assays.

Catalog No. 50-002-36405 Supplier Medchemexpress LLC Supplier No. HY12579110MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

cis-Epoxysuccinic acid is a small-molecule agonist of the succinate receptor (SUCNR1) used in receptor pharmacology, cell signaling research, and biochemical assays. It is supplied as a white to off-white solid and as a solution (commonly 10 mM in DMSO, 1 mL); handle according to the safety data sheet and storage recommendations.

  • Purity 98.0%.
  • Molecular weight 132.07 g/mol.
  • Chemical formula C4H4O5.
  • Available as 10 mM solution in DMSO (1 mL) and solid sizes 100 mg-5 g.
  • Appearance white to off-white solid.
  • Intended for SUCNR1 pharmacology and biochemical assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.