Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cis isoeugenol | 5912-86-7 | 95.0% | 164.20 g/mol | C10H12O2 | 10 MG
SDP

Catalog No. 5000295587
Encompass_Preferred

cis-Isoeugenol is the cis stereoisomer of isoeugenol, provided as a research-grade small molecule for analytical, antibacterial, and fragrance-related studies. It is offered in small sample sizes for laboratory use and is characterized by a defined purity and storage profile.

  • High purity: 95.0%.
  • Chemical formula: C10H12O2.
  • Molecular weight: 164.20 g/mol.
  • CAS number: 5912-86-7.
  • Suggested storage: pure form -20°C; in solvent -80°C for 6 months or -20°C for 1 month.
  • Common applications: antibacterial assays, fragrance research, and analytical reference work.

Catalog No. 50-002-95587 Supplier Medchemexpress LLC Supplier No. HYN1952A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

cis-Isoeugenol is the cis stereoisomer of isoeugenol, provided as a research-grade small molecule for analytical, antibacterial, and fragrance-related studies. It is offered in small sample sizes for laboratory use and is characterized by a defined purity and storage profile.

  • High purity: 95.0%.
  • Chemical formula: C10H12O2.
  • Molecular weight: 164.20 g/mol.
  • CAS number: 5912-86-7.
  • Suggested storage: pure form -20°C; in solvent -80°C for 6 months or -20°C for 1 month.
  • Common applications: antibacterial assays, fragrance research, and analytical reference work.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.