Learn More
Medchemexpress LLC Cis-Jasmone | 488-10-8 | 500 MG
SDP
Catalog No. 5000419619
Encompass_Preferred
Cis-Jasmone is a plant-derived natural product. It is released by many flowers and sometimes by leaves to attract pollinators or as a chemical cue for host location by insect flower herbivores. Treating crop plants with cis-Jasmone not only provides direct defense against herbivores but also induces indirect defense by releasing volatile organic compounds (VOCs) that attract natural enemies.
  • Induces direct defense against herbivores
  • Induces indirect defense by releasing volatile organic compounds that attract natural enemies
  • Exhibits a repellent effect on aphids by activating olfactory receptors
  • Attracts aphid natural enemies through the release of plant volatile substances
Catalog No. 50-004-19619 Supplier Medchemexpress LLC Supplier No. HYN7058500MG
Cis-Jasmone is a plant-derived natural product. It is released by many flowers and sometimes by leaves to attract pollinators or as a chemical cue for host location by insect flower herbivores. Treating crop plants with cis-Jasmone not only provides direct defense against herbivores but also induces indirect defense by releasing volatile organic compounds (VOCs) that attract natural enemies.
  • Induces direct defense against herbivores
  • Induces indirect defense by releasing volatile organic compounds that attract natural enemies
  • Exhibits a repellent effect on aphids by activating olfactory receptors
  • Attracts aphid natural enemies through the release of plant volatile substances