Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Organic Chemistry cis-Limonene oxide | 25G | 4680-24-4 | MFCD09996820

Catalog No. 90626325G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass

cis-Limonene oxide , 25G

About this item:

CAS #: 4680-24-4
MDL #: MFCD09996820
Molecular Weight: 152.23
UNSPSC Code: 12352200

Catalog No. 90-626-325G Supplier Sigma Organic Chemistry Supplier No. 90626325G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

cis-Limonene oxide , 25G

About this item:

CAS #: 4680-24-4
MDL #: MFCD09996820
Molecular Weight: 152.23
UNSPSC Code: 12352200

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.