Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (-)-Cis-Myrtanylamine

Catalog No. 18080710G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-080-710G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 18-080-710G Supplier Sigma Aldrich Supplier No. 18080710G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(-)-cis-Myrtanylamine may be used:As a reactant to prepare N,N-bis((-)-cis-myrtanyl)butylene-2,3-diimine (BMDI), a bidentate diimine ligand that can form transition metal complexes for catalyzing asymmetric synthesis.As a building block to prepare imidazole derivatives, which are potent and selective cannabinoid receptor (CB2) antagonists.To prepare a chiral catalyst, which can catalyze asymmetric three-component Mannich reaction.

Specifications

CAS 42856-71-3
Molecular Formula C6H11NO2
Refractive Index n20/D 1.4877
Linear Formula C6H11NO2
Quantity 250 mg
MDL Number MFCD01318647
Molecular Weight (g/mol) 129.16 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.