Learn More
TARGETMOL CHEMICALS INC CIS-N-FERULOYLTYRAMINE 5MG
SDP
Catalog No. 502578873
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Cis-N-Feruloyltyramine is a naturally occurring compound found in various plants that shows cytotoxicity against the P-388 cancer cell line. Cis-N-Feruloyltyramine is an inhibitor of in vitro prostaglandin (PG) synthesis. purity: 98%
Catalog No. 50-257-8873 Supplier TARGETMOL CHEMICALS INC Supplier No. TN36695MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Cis-N-Feruloyltyramine is a naturally occurring compound found in various plants that shows cytotoxicity against the P-388 cancer cell line. Cis-N-Feruloyltyramine is an inhibitor of in vitro prostaglandin (PG) synthesis. purity: 98%