Learn More
Medchemexpress LLC cis-p-Coumaric acid | 4501-31-9 | 164.16 | 1 MG
SDP
Catalog No. 5000389304
Encompass_Preferred
cis-p-Coumaric acid, also known as cis-4-Hydroxycinnamic acid, is the cis-isomer of p-Coumaric acid. This compound is notably found in higher concentrations in viable groundnut seeds and exhibits oral activity. It plays a role in cellular processes, including the inhibition of cell proliferation and the promotion of apoptosis.
  • Exhibits oral activity
  • Inhibits cell proliferation
  • Promotes apoptosis
  • Shows neuroprotective activity against glutamate-induced cell death
  • White to off-white solid appearance
  • Molecular formula C9H8O3
Catalog No. 50-003-89304 Supplier Medchemexpress LLC Supplier No. HYN0351A1MG
cis-p-Coumaric acid, also known as cis-4-Hydroxycinnamic acid, is the cis-isomer of p-Coumaric acid. This compound is notably found in higher concentrations in viable groundnut seeds and exhibits oral activity. It plays a role in cellular processes, including the inhibition of cell proliferation and the promotion of apoptosis.
  • Exhibits oral activity
  • Inhibits cell proliferation
  • Promotes apoptosis
  • Shows neuroprotective activity against glutamate-induced cell death
  • White to off-white solid appearance
  • Molecular formula C9H8O3