Learn More
Cayman Chemical cis-Petroselinic Acid, Purity: ≥98%, CAS: 593-39-5, 50 mg
SDP
Catalog No. 502982957
Encompass_Preferred
A monounsaturated fatty acid and isomer of oleic acid; major component of plant lipids; rats fed a diet containing it have a reduction in arachidonic acid levels and an increase in linoleic acid levels in the heart, liver, and blood; has been used in a composite membrane to study plant uptake; has been used in the synthesis of new biosurfactants (sophorolipids)
Catalog No. 50-298-2957 Supplier Cayman Chemical Supplier No. 2002450 MG
A monounsaturated fatty acid and isomer of oleic acid; major component of plant lipids; rats fed a diet containing it have a reduction in arachidonic acid levels and an increase in linoleic acid levels in the heart, liver, and blood; has been used in a composite membrane to study plant uptake; has been used in the synthesis of new biosurfactants (sophorolipids)