Learn More
Medchemexpress LLC Cis-piperidine-2,3-dicarboxylic acid | 46026-75-9 | 98.0% | 100 MG
SDP
Catalog No. 5000231240
Encompass_Preferred
Cis-piperidine-2,3-dicarboxylic acid is an ionotropic glutamate receptor ligand used in neuroscience research. It functions as a non-specific antagonist of NMDA, AMPA, and kainate receptors and as a partial agonist at NMDA receptors, making it useful for studying excitatory synaptic transmission.
  • Acts as a non-specific antagonist of NMDA, AMPA, and kainate receptors.
  • Displays partial agonist activity at NMDA receptors for mechanistic studies.
  • Provided in small milligram package sizes suitable for laboratory research.
  • Purity typically reported as 98.0%.
Catalog No. 50-002-31240 Supplier Medchemexpress LLC Supplier No. HY121964100MG
Cis-piperidine-2,3-dicarboxylic acid is an ionotropic glutamate receptor ligand used in neuroscience research. It functions as a non-specific antagonist of NMDA, AMPA, and kainate receptors and as a partial agonist at NMDA receptors, making it useful for studying excitatory synaptic transmission.
  • Acts as a non-specific antagonist of NMDA, AMPA, and kainate receptors.
  • Displays partial agonist activity at NMDA receptors for mechanistic studies.
  • Provided in small milligram package sizes suitable for laboratory research.
  • Purity typically reported as 98.0%.