Learn More
Medchemexpress LLC Cis-piperidine-2,3-dicarboxylic acid | 46026-75-9 | >98.0% | 50 MG
SDP
Catalog No. 5000231130
Encompass_Preferred
Cis-piperidine-2,3-dicarboxylic acid is a research reagent that modulates ionotropic glutamate receptors. It functions as a non-specific antagonist of NMDA, AMPA, and kainate receptors and as a partial agonist at NMDA receptors, and is used to block or study excitatory synaptic transmission in neuropharmacology experiments.
  • Acts as a non-specific antagonist of NMDA, AMPA, and kainate receptors.
  • Exhibits partial agonist activity at NMDA receptors for modulation studies.
  • High purity (≥98.0%) suitable for research applications.
  • Soluble in water (≈100 mg/mL) with ultrasonic assistance.
  • Off-white to light yellow solid stored at 4°C and protected from light.
Catalog No. 50-002-31130 Supplier Medchemexpress LLC Supplier No. HY12196450MG
Cis-piperidine-2,3-dicarboxylic acid is a research reagent that modulates ionotropic glutamate receptors. It functions as a non-specific antagonist of NMDA, AMPA, and kainate receptors and as a partial agonist at NMDA receptors, and is used to block or study excitatory synaptic transmission in neuropharmacology experiments.
  • Acts as a non-specific antagonist of NMDA, AMPA, and kainate receptors.
  • Exhibits partial agonist activity at NMDA receptors for modulation studies.
  • High purity (≥98.0%) suitable for research applications.
  • Soluble in water (≈100 mg/mL) with ultrasonic assistance.
  • Off-white to light yellow solid stored at 4°C and protected from light.