Learn More
Abcam cis-PPDA, GluN2C/GluN2D NMDA antagonist, 50MG
Catalog No. 502707899
Encompass_Preferred
MW 378.4 Da, Purity >98%. Potent GluN2C/GluN2D (formerly NR2C / NR2D)-preferring NMDA receptor antagonist. (Ki values for recombinant rat receptors are 0.096 (NR2C), 0.125 (NR2D), 0.55 (NR2A) and 0.31 μM (NR2B)).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7899 Supplier Abcam Supplier No. AB12004750MG
MW 378.4 Da, Purity >98%. Potent GluN2C/GluN2D (formerly NR2C / NR2D)-preferring NMDA receptor antagonist. (Ki values for recombinant rat receptors are 0.096 (NR2C), 0.125 (NR2D), 0.55 (NR2A) and 0.31 μM (NR2B)).
The product is subject to the following: Abcam Restricted Use Statement