Learn More
Medchemexpress LLC (±)-cis-Sertraline-d3 hydrochloride | 1217741-83-7 | 99.5% | 5 MG
SDP
Catalog No. 5000406248
Encompass_Preferred
(±)-cis-Sertraline-d3 (hydrochloride) is the deuterium labeled Sertraline hydrochloride, an antidepressant belonging to the selective serotonin reuptake inhibitor (SSRI) class. It is researched for various diseases, including major depressive disorder and obsessive.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-06248 Supplier Medchemexpress LLC Supplier No. HYB0176AS15MG
(±)-cis-Sertraline-d3 (hydrochloride) is the deuterium labeled Sertraline hydrochloride, an antidepressant belonging to the selective serotonin reuptake inhibitor (SSRI) class. It is researched for various diseases, including major depressive disorder and obsessive.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS