Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich cis-Stilbene

Catalog No. S480810G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S480810G 10 g
S48085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S480810G Supplier Sigma Aldrich Supplier No. S480810G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

cis-Stilbene can undergo epoxidation in the presence of supported gold nanoparticles (AuNP) and using cumene as a solvent to form a mixture of cis- and trans-stilbene oxides.

Specifications

CAS 645-49-8
Density 1.011 g/mL (at 25°C (literature))
Boiling Point 82°C to 84°C (0.4 mmHg)
Molecular Formula C14H12
Refractive Index n20/D 1.622 (literature)
Linear Formula C6H5CH=CHC6H5
MDL Number MFCD00004788
Quantity 10 g
Synonym cis-1,2-Diphenylethylene; Isostilbene
Molecular Weight (g/mol) 180.25
Percent Purity 96%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.