Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-Stilbene 97%, Thermo Scientific™

Catalog No. AC132770100
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC132770100 10 g
AC132770050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC132770100 Supplier Thermo Scientific Chemicals Supplier No. 132770100

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 645-49-8
Molecular Formula C14H12
Molecular Weight (g/mol) 180.25
InChI Key PJANXHGTPQOBST-QXMHVHEDSA-N
Synonym cis-stilbene, z-stilbene, z-1,2-diphenylethene, cis-1,2-diphenylethylene, isostilbene, cis-diphenylethene, stilbene, z, z-2-phenylethenyl benzene, benzene, 1,1'-1,2-ethenediyl bis-, z, z-1,2-diphenylethylene
PubChem CID 5356785
ChEBI CHEBI:36008
IUPAC Name (Z)-stilbene
SMILES C1=CC=C(C=C1)C=CC2=CC=CC=C2

Specifications

Color Colorless to Yellow
Density 1.01
Infrared Spectrum Authentic
Linear Formula C6H5CH=CHC6H5
Refractive Index 1.6210 to 1.6230 (20°C, 589nm)
Quantity 10 g
Beilstein 05, 630
Merck Index 15, 8946
Formula Weight 180.25
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material cis-Stilbene, 97%
Show More Show Less
EINECSNumber 211-445-7
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.