Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis,trans-5,9-Cyclododecadiene-cis-1,2-diol 98.0+%, TCI America™
SDP

Catalog No. C03555G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C03555G 5 g
C035525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C03555G Supplier TCI America Supplier No. C03555G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 29118-70-5
Molecular Formula C12H20O2
Molecular Weight (g/mol) 196.29
MDL Number MFCD00021191
InChI Key BHYILZSOXKSYCF-YZXZKFOESA-N
Synonym cis-1,2-Dihydroxy-cis,trans-5,9-cyclododecadiene
PubChem CID 133126446
IUPAC Name (2S,5E,9E)-cyclododeca-5,9-diene-1,2-diol
SMILES C1CC=CCCC(C(CCC=C1)O)O

Specifications

Melting Point 159°C
Quantity 5 g
Formula Weight 196.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material cis,trans-5,9-Cyclododecadiene-cis-1,2-diol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.